Anti-NR2E3 Mouse Monoclonal Antibody [clone: 3G3]

Leverancier: OriGene
CF806218
ORIGCF806218EA 764 EUR
ORIGCF806218
Anti-NR2E3 Mouse Monoclonal Antibody [clone: 3G3]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NR2E3
Clonality: Monoclonal
Clone: 3G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR