Anti-RARA Mouse Monoclonal Antibody [clone: 2D2]

Leverancier: OriGene
CF806244
ORIGCF806244EA 764 EUR
ORIGCF806244
Anti-RARA Mouse Monoclonal Antibody [clone: 2D2]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RARA
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR