Anti-TGFBR1 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA323850EA 514 EUR
ORIGTA323850
Anti-TGFBR1 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:3000-10000; Immunochemistry: 1:50-200

Type: Primary
Antigen: TGFBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR