Anti-HTR3A Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA323541EA 564 EUR
ORIGTA323541
Anti-HTR3A Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:1000-2000; Western Blot: 1:200-1000; Immunochemistry: 1:25-100

Type: Primary
Antigen: HTR3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR