Anti-UCHL1 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA327726EA 1230 EUR
ORIGTA327726
Anti-UCHL1 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR