Anti-ZAP70 Mouse Monoclonal Antibody [clone: 2F3.2]

Leverancier: OriGene
ORIGTA327755EA 1020 EUR
ORIGTA327755
Anti-ZAP70 Mouse Monoclonal Antibody [clone: 2F3.2]
Antilichamen
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: 2F3.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR