Anti-GLP1R Goat Polyclonal Antibody

Leverancier: OriGene
ORIGTA326308EA 500 EUR
ORIGTA326308
Anti-GLP1R Goat Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: GLP1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR