Anti-AHR Goat Polyclonal Antibody

Leverancier: OriGene
ORIGTA326329EA 502 EUR
ORIGTA326329
Anti-AHR Goat Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: AHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR