Anti-MAPK14 Rabbit Polyclonal Antibody [clone: N/A]

Leverancier: OriGene
ORIGTA326350EA 484 EUR
ORIGTA326350
Anti-MAPK14 Rabbit Polyclonal Antibody [clone: N/A]
Antilichamen
Recommended Dilutions: Western Blot: 1:5000; Immunoprecipitation: 1:250

Type: Primary
Antigen: MAPK14
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, canine, Chicken, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR