Anti-HSPA1A Mouse Monoclonal Antibody [clone: C92F3A-5]

Leverancier: OriGene
ORIGTA326356EA 669 EUR
ORIGTA326356
Anti-HSPA1A Mouse Monoclonal Antibody [clone: C92F3A-5]
Antilichamen
Recommended Dilutions: 1?g/ml

Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: C92F3A-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Bovine, C.elegans, canine, carp, Chicken, drosophilia, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR