Anti-HSPA1A Mouse Monoclonal Antibody [clone: 2A4]

Leverancier: OriGene
ORIGTA326359EA 596 EUR
ORIGTA326359
Anti-HSPA1A Mouse Monoclonal Antibody [clone: 2A4]
Antilichamen
Recommended Dilutions: Immunofluorescence: 1:100; Immunochemistry: 1:100; Western Blot: 1:5000

Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: 2A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: amphibian, Chicken, Fish, Drosophila, Human, Mouse, Rat, saccharomyces cerevisiae
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR