Anti-HSPA8 Mouse Monoclonal Antibody [clone: 1F2-H5]

Leverancier: OriGene
ORIGTA326362EA 681 EUR
ORIGTA326362
Anti-HSPA8 Mouse Monoclonal Antibody [clone: 1F2-H5]
Antilichamen
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HSPA8
Clonality: Monoclonal
Clone: 1F2-H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat.
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR