Anti-HSPA8 Rabbit Polyclonal Antibody [clone: N/A]

Leverancier: OriGene
ORIGTA326363EA 575 EUR
ORIGTA326363
Anti-HSPA8 Rabbit Polyclonal Antibody [clone: N/A]
Antilichamen
Recommended Dilutions: Western Blot: 1:1500-2000

Type: Primary
Antigen: HSPA8
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster, Human, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR