Anti-HSPD1 Rabbit Polyclonal Antibody [clone: N/A]
Leverancier: OriGene
ORIGTA326375EA 636 EUR
ORIGTA326375
Anti-HSPD1 Rabbit Polyclonal Antibody [clone: N/A]
Antilichamen
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: HSPD1
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, canine, Chicken, Human, Mouse, Rabbit, Rat