Anti-TRAP1 Mouse Monoclonal Antibody [clone: Trap1-6]

Leverancier: OriGene
ORIGTA326389EA 591 EUR
ORIGTA326389
Anti-TRAP1 Mouse Monoclonal Antibody [clone: Trap1-6]
Antilichamen
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TRAP1
Clonality: Monoclonal
Clone: Trap1-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR