Anti-KCNQ4 Mouse Monoclonal Antibody [clone: S43-6]

Leverancier: OriGene
ORIGTA326401EA 632 EUR
ORIGTA326401
Anti-KCNQ4 Mouse Monoclonal Antibody [clone: S43-6]
Antilichamen
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: KCNQ4
Clonality: Monoclonal
Clone: S43-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR