Anti-PCLO Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA326449EA 632 EUR
ORIGTA326449
Anti-PCLO Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PCLO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR