Anti-KIF11 Mouse Monoclonal Antibody [clone: 10C7/Eg5]

Leverancier: OriGene
ORIGTA327996EA 536 EUR
ORIGTA327996
Anti-KIF11 Mouse Monoclonal Antibody [clone: 10C7/Eg5]
Antilichamen
Type: Primary
Antigen: KIF11
Clonality: Monoclonal
Clone: 10C7/Eg5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR