Anti-HCN3 Mouse Monoclonal Antibody [clone: S141-28]

Leverancier: OriGene
ORIGTA326475EA 574 EUR
ORIGTA326475
Anti-HCN3 Mouse Monoclonal Antibody [clone: S141-28]
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: HCN3
Clonality: Monoclonal
Clone: S141-28
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR