Anti-KCNA2 Mouse Monoclonal Antibody [clone: S14-16]

Leverancier: OriGene
ORIGTA326482EA 632 EUR
ORIGTA326482
Anti-KCNA2 Mouse Monoclonal Antibody [clone: S14-16]
Antilichamen
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml

Type: Primary
Antigen: KCNA2
Clonality: Monoclonal
Clone: S14-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Xenopus, zebraFish
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR