Anti-Tcp1 Rat Monoclonal Antibody [clone: 23c]

Leverancier: OriGene
ORIGTA326506EA 591 EUR
ORIGTA326506
Anti-Tcp1 Rat Monoclonal Antibody [clone: 23c]
Antilichamen
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Tcp1
Clonality: Monoclonal
Clone: 23c
Conjugation: Unconjugated
Epitope: Full length
Host: Rat
Isotype: IgG2c
Reactivity: Bovine, Dog, Hamster, Mouse, Rabbit, Rat, Sheep
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR