Anti-Grin2a Mouse Monoclonal Antibody [clone: S327-95]
Leverancier: OriGene
ORIGTA326537EA 692 EUR
ORIGTA326537
Anti-Grin2a Mouse Monoclonal Antibody [clone: S327-95]
Antilichamen
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: Grin2a
Clonality: Monoclonal
Clone: S327-95
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2A
Reactivity: Human, Mouse, Rat