Anti-SSRP1 Mouse Monoclonal Antibody [clone: 10D1]

Leverancier: OriGene
ORIGTA327961EA 469 EUR
ORIGTA327961
Anti-SSRP1 Mouse Monoclonal Antibody [clone: 10D1]
Antilichamen
Type: Primary
Antigen: SSRP1
Clonality: Monoclonal
Clone: 10D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR