Anti-EIF3D Mouse Monoclonal Antibody [clone: 8A9]

Leverancier: OriGene
ORIGTA327977EA 466 EUR
ORIGTA327977
Anti-EIF3D Mouse Monoclonal Antibody [clone: 8A9]
Type: Primary
Antigen: EIF3D
Clonality: Monoclonal
Clone: 8A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR