Anti-Kcnb1 Mouse Monoclonal Antibody [clone: S39-25]

Leverancier: OriGene
ORIGTA326554EA 632 EUR
ORIGTA326554
Anti-Kcnb1 Mouse Monoclonal Antibody [clone: S39-25]
Antilichamen
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml

Type: Primary
Antigen: Kcnb1
Clonality: Monoclonal
Clone: S39-25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR