Anti-FOXP3 Rabbit Polyclonal Antibody [clone: Poly6238]

Leverancier: OriGene
ORIGTA328027EA 466 EUR
ORIGTA328027
Anti-FOXP3 Rabbit Polyclonal Antibody [clone: Poly6238]
Type: Primary
Antigen: FOXP3
Clonality: Polyclonal
Clone: Poly6238
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR