Anti-BCL11B Rat Monoclonal Antibody [clone: 25B6]

Leverancier: OriGene
ORIGTA328046EA 442 EUR
ORIGTA328046
Anti-BCL11B Rat Monoclonal Antibody [clone: 25B6]
Antilichamen
Type: Primary
Antigen: BCL11B
Clonality: Monoclonal
Clone: 25B6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Human, Mouse, zebraFish
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR