Anti-KPNA7 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA326608EA 557 EUR
ORIGTA326608
Anti-KPNA7 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: KPNA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR