Anti-FCHO2 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA326623EA 557 EUR
ORIGTA326623
Anti-FCHO2 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 2.5 ?g/ml

Type: Primary
Antigen: FCHO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR