Anti-MAP1LC3A Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA326682EA 564 EUR
ORIGTA326682
Anti-MAP1LC3A Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: MAP1LC3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR