Anti-CCR7 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA326705EA 564 EUR
ORIGTA326705
Anti-CCR7 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: CCR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR