Anti-NTRK2 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA325756EA 514 EUR
ORIGTA325756
Anti-NTRK2 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: NTRK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR