Anti-KRT19 Goat Polyclonal Antibody

Leverancier: OriGene
ORIGTA326741EA 547 EUR
ORIGTA326741
Anti-KRT19 Goat Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.03-0.1µg/ml

Type: Primary
Antigen: KRT19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR