Anti-CTDSP1 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA326105EA 514 EUR
ORIGTA326105
Anti-CTDSP1 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: CTDSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR