Anti-TAOK3 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA326132EA 564 EUR
ORIGTA326132
Anti-TAOK3 Rabbit Polyclonal Antibody
Antilichamen
  • Recommended Dilutions: Western Blot: 1:500-1:3000 ELISA: 1:10000

Type: Primary
Antigen: TAOK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR