Anti-Glp1r Goat Polyclonal Antibody

Leverancier: OriGene
ORIGTA326758EA 500 EUR
ORIGTA326758
Anti-Glp1r Goat Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:128000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: Glp1r
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR