Anti-CD1A Rabbit Monoclonal Antibody [clone: EP3622]

Leverancier: OriGene
ORIGTA327624EA 1270 EUR
ORIGTA327624
Anti-CD1A Rabbit Monoclonal Antibody [clone: EP3622]
Antilichamen
Recommended Dilutions: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: EP3622
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR