Anti-CR2 Rabbit Monoclonal Antibody [clone: EP3093]

Leverancier: OriGene
ORIGTA327628EA 1060 EUR
ORIGTA327628
Anti-CR2 Rabbit Monoclonal Antibody [clone: EP3093]
Antilichamen
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: EP3093
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR