Anti-FCER2 Mouse Monoclonal Antibody [clone: 1B12]

Leverancier: OriGene
ORIGTA327629EA 1520 EUR
ORIGTA327629
Anti-FCER2 Mouse Monoclonal Antibody [clone: 1B12]
Antilichamen
Recommended Dilutions: Immunochemistry: 1:10-1:50

Type: Primary
Antigen: FCER2
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR