Anti-PTPRC Mouse Monoclonal Antibody [clone: 2B11 & PD7/26]

Leverancier: OriGene
ORIGTA327641EA 513 EUR
ORIGTA327641
Anti-PTPRC Mouse Monoclonal Antibody [clone: 2B11 & PD7/26]
Antilichamen
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 2B11 & PD7/26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR