Anti-CD5 Mouse Monoclonal Antibody [clone: 4C7]

Leverancier: OriGene
ORIGTA327644EA 1430 EUR
ORIGTA327644
Anti-CD5 Mouse Monoclonal Antibody [clone: 4C7]
Antilichamen
Recommended Dilutions: Immunochemistry: 1:10-1:50

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 4C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR