Anti-EPCAM Mouse Monoclonal Antibody [clone: Ber-EP4]

Leverancier: OriGene
ORIGTA327674EA 1020 EUR
ORIGTA327674
Anti-EPCAM Mouse Monoclonal Antibody [clone: Ber-EP4]
Antilichamen
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: Ber-EP4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR