Anti-MUC5AC Mouse Monoclonal Antibody [clone: MRQ-19]

Leverancier: OriGene
ORIGTA327703EA 746 EUR
ORIGTA327703
Anti-MUC5AC Mouse Monoclonal Antibody [clone: MRQ-19]
Antilichamen
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: MUC5AC
Clonality: Monoclonal
Clone: MRQ-19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR