Anti-NGFR Mouse Monoclonal Antibody [clone: MRQ-21]

Leverancier: OriGene
ORIGTA327714EA 1520 EUR
ORIGTA327714
Anti-NGFR Mouse Monoclonal Antibody [clone: MRQ-21]
Antilichamen
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: NGFR
Clonality: Monoclonal
Clone: MRQ-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR