Anti-PARP4 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA326252EA 514 EUR
ORIGTA326252
Anti-PARP4 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100ELISA: 1:10000

Type: Primary
Antigen: PARP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR