Anti-TRAF2 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA326864EA 513 EUR
ORIGTA326864
Anti-TRAF2 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:20-1:200; Immunofluorescence: 1:20-1:100; Immunoprecipitation: 1:20-1:100

Type: Primary
Antigen: TRAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR