Anti-CEACAM5 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA326869EA 513 EUR
ORIGTA326869
Anti-CEACAM5 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: CEACAM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR