Anti-NTSR1 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA328679EA 952 EUR
ORIGTA328679
Anti-NTSR1 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:200-1:2000; Immunochemistry: 1:100-1:3000; Flow Cytometry: 1:50-1:600;

Type: Primary
Antigen: NTSR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR