Anti-ZDHHC18 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA329493EA 518 EUR
ORIGTA329493
Anti-ZDHHC18 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ZDHHC18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR