Anti-FN1 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA590325EA 625 EUR
ORIGTA590325
Anti-FN1 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:100-1:2000

Type: Primary
Antigen: FN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR